12H-Benzofuro[3,2-a]carbazole 98%

Supplier: Ambeed
Warning

A1328738-1G A1328738-250MG A1328738-5G
76681-904EA 44.56 USD
76681-904 76681-906 76681-908
12H-Benzofuro[3,2-a]carbazole 98%
12H-Benzofuro[3,2-a]carbazole

Precaución: for Research Use Only

Formula: C₁₈H₁₁NO
MW: 257.2860 g/mol
CAS Number: 1246308-85-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR