2,2'-Bipyrazine 99+%

Supplier: Ambeed
Warning

A1328585-100MG A1328585-1G A1328585-250MG A1328585-5G
76681-894EA 28 USD
76681-894 76681-896 76681-898 76681-900
2,2'-Bipyrazine 99+%
2,2'-Bipyrazine

Precaución: for Research Use Only

Formula: C₈H₆N₄
MDL Number: MFCD00010288
CAS Number: 10199-00-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR