Tetrahydroxy-1,4-benzoquinone hydrate 97%

Supplier: Ambeed
Warning

Synonyms: Tetrahydroxyquinone, 2,3,5,6-Tetrahydroxycyclohexa-2,5-diene-1,4-dione xhydrate, Tetroquinone

A1328533-1G A1328533-5G
76681-888EA 50 USD
76681-888 77621-552
Tetrahydroxy-1,4-benzoquinone hydrate 97%
Tetrahydroxy-1,4-benzoquinone Hydrate

Precaución: for Research Use Only

Formula: C₆H₆O₇
MW: 190.11 g/mol
MDL Number: MFCD00001597
CAS Number: 123334-16-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR