(1S)-(-)-Camphanic chloride 95%, (Chunks or pellets)

Supplier: Ambeed
Danger

Synonyms: (1S)-4,7,7-Trimethyl-3-oxo-2-oxabicyclo[2.2.1]heptane-1-carbonyl chloride, (-)-Camphanic acid chloride

A1328343-1G A1328343-25G A1328343-5G
76681-882EA 9.55 USD
76681-882 76681-884 76681-886
(1S)-(-)-Camphanic chloride 95%, (Chunks or pellets)
(1S)-(-)-Camphanic chloride

Precaución: for Research Use Only

Formula: C₁₀H₁₃ClO₃
MW: 216.67 g/mol
MDL Number: MFCD00135626
CAS Number: 39637-74-6
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR