(S)-2-(2-(Bis(3,5-bis(trifluoromethyl)phenyl)phosphino)phenyl)-4-(tert-butyl)-4,5-dihydrooxazole 98%

Supplier: Ambeed
Warning

A1327715-100MG A1327715-1G A1327715-250MG
76681-868EA 129 USD
76681-868 77867-310 77867-311
(S)-2-(2-(Bis(3,5-bis(trifluoromethyl)phenyl)phosphino)phenyl)-4-(tert-butyl)-4,5-dihydrooxazole 98%
(S)-2-(2-(Bis(3,5-bis(trifluoromethyl)phenyl)phosphino)phenyl)-4-(tert-butyl)-4,5-dihydrooxazole

Precaución: for Research Use Only

Formula: C₂₉H₂₂F₁₂NOP
CAS Number: 1092491-37-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR