6-Bromo-3-(2-chloro-5-(trifluoromethyl)pyrimidin-4-yl)-1-(phenylsulfonyl)-1H-indole 98%

Supplier: Ambeed
Warning

A1327434-250MG A1327434-100MG
76681-866EA 14 USD
76681-866 77867-304
6-Bromo-3-(2-chloro-5-(trifluoromethyl)pyrimidin-4-yl)-1-(phenylsulfonyl)-1H-indole 98%
6-Bromo-3-(2-chloro-5-(trifluoromethyl)pyrimidin-4-yl)-1-(phenylsulfonyl)-1H-indole

Precaución: for Research Use Only

Formula: C₁₉H₁₀BrClF₃N₃O₂S
MDL Number: MFCD32640880
CAS Number: 2173193-44-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR