tert-Butyl 3-(2-iodoethoxy)propanoate 98%

Supplier: Ambeed
Warning

A1327425-100MG A1327425-1G A1327425-250MG A1327425-5G
76681-858EA 24.41 USD
76681-858 76681-860 76681-862 76681-864
tert-Butyl 3-(2-iodoethoxy)propanoate 98%
tert-Butyl 3-(2-iodoethoxy)propanoate

Precaución: for Research Use Only

Formula: C₉H₁₇IO₃
MW: 300.1340 g/mol
MDL Number: MFCD32689526
CAS Number: 2296723-16-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR