5,5'-Methylenediisophthalic acid 98%

Supplier: Ambeed
Warning

A1327396-100MG A1327396-250MG A1327396-5G A1327396-1G
76681-852EA 29 USD
76681-852 76681-854 76681-856 77566-584
5,5'-Methylenediisophthalic acid 98%
5,5'-Methylenediisophthalic acid

Precaución: For research use only.

Formula: C₁₇H₁₂O₈
MDL Number: MFCD19053170
CAS Number: 10397-52-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR