(S)-2,5-Dimethoxy-N-(quinuclidin-3-yl)benzamide 2,2,2-trifluoroacetate 98%

Supplier: Ambeed
Warning

A1327316-10MG A1327316-1MG A1327316-25MG A1327316-50MG A1327316-5MG A1327316-250MG
76681-842EA 59 USD
76681-842 76681-844 76681-846 76681-848 76681-850 77867-297
(S)-2,5-Dimethoxy-N-(quinuclidin-3-yl)benzamide 2,2,2-trifluoroacetate 98%
(S)-2,5-Dimethoxy-N-(quinuclidin-3-yl)benzamide 2,2,2-trifluoroacetate

Precaución: for Research Use Only

Formula: C₁₈H₂₃F₃N₂O₅
MDL Number: MFCD31689251
CAS Number: 1336913-03-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR