4-Iodo-2-((tosyloxy)methyl)phenyl 4-methylbenzenesulfonate 97%

Supplier: Ambeed
Warning

A1327302-100MG A1327302-1G A1327302-250MG
76681-836EA 9 USD
76681-836 76681-838 76681-840
4-Iodo-2-((tosyloxy)methyl)phenyl 4-methylbenzenesulfonate 97%
4-Iodo-2-((tosyloxy)methyl)phenyl 4-methylbenzenesulfonate

Precaución: for Research Use Only

Formula: C₂₁H₁₉IO₆S₂
MDL Number: MFCD32856771
CAS Number: 2682115-56-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR