9,9-Didodecyl-9H-fluorene-2,7-dicarbaldehyde 95%

Supplier: Ambeed
Warning

A1326911-1G A1326911-250MG A1326911-5G
76681-814EA 88.06 USD
76681-814 76681-816 76681-818
9,9-Didodecyl-9H-fluorene-2,7-dicarbaldehyde 95%
9,9-Didodecyl-9H-fluorene-2,7-dicarbaldehyde

Precaución: for Research Use Only

Formula: C₃₉H₅₈O₂
MW: 558.8766 g/mol
MDL Number: MFCD32641195
CAS Number: 1450727-06-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR