(4-Fluoro-2-(1H-imidazol-1-yl)phenyl)methanamine 97%

Supplier: Ambeed
Warning

A1326649-1G A1326649-250MG
76681-810EA 187.78 USD
76681-810 76681-812
(4-Fluoro-2-(1H-imidazol-1-yl)phenyl)methanamine 97%
(4-Fluoro-2-(1H-imidazol-1-yl)phenyl)methanamine

Precaución: for Research Use Only

Formula: C₁₀H₁₀FN₃
MW: 191.2049 g/mol
MDL Number: MFCD28523199
CAS Number: 869591-89-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR