(2S,4R)-1-((S)-2-(3-(3-((S)-2-(((7-(8-Chloronaphthalen-1-yl)-4-((S)-3-(cyanomethyl)-4-(2-fluoroacryloyl)piperazin-1-yl)-5,6,7,8-tetrahydropyrido[3,4-d]pyrimidin-2-yl)oxy)methyl)pyrrolidin-1-yl)propoxy)propanamido)-3,3-dimethylbutanoyl)-4-hydroxy-N-(4-(4-methylthiazol-5-yl)benzyl)pyrrolidine-2-carboxamide 98%

Supplier: Ambeed
Warning

Synonyms: LC-2, (2S,4R)-1-((S)-2-(3-(3-((S)-2-(((7-(8-Chloronaphthalen-1-yl)-4-((S)-3-(cyanomethyl)-4-(2-fluoroacryloyl)piperazin-1-yl)-5,6,7,8-tetrahydropyrido[3,4-d]pyrimidin-2-yl)oxy)methyl)pyrrolidin-1-yl)propoxy)propanamido)-3,3-dimethylbutanoyl)-4-hydroxy-N-(4-(4-methylthiazol-5-yl)benzyl)pyrrolidine-2-carboxamide

A1325961-10MG A1325961-1MG A1325961-25MG A1325961-50MG
76681-782EA 381.93 USD
76681-782 76681-784 76681-786 77621-508
(2S,4R)-1-((S)-2-(3-(3-((S)-2-(((7-(8-Chloronaphthalen-1-yl)-4-((S)-3-(cyanomethyl)-4-(2-fluoroacryloyl)piperazin-1-yl)-5,6,7,8-tetrahydropyrido[3,4-d]pyrimidin-2-yl)oxy)methyl)pyrrolidin-1-yl)propoxy)propanamido)-3,3-dimethylbutanoyl)-4-hydroxy-N-(4-(4-methylthiazol-5-yl)benzyl)pyrrolidine-2-carboxamide 98%
(2S,4R)-1-((S)-2-(3-(3-((S)-2-(((7-(8-Chloronaphthalen-1-yl)-4-((S)-3-(cyanomethyl)-4-(2-fluoroacryloyl)piperazin-1-yl)-5,6,7,8-tetrahydropyrido[3,4-d]pyrimidin-2-yl)oxy)methyl)pyrrolidin-1-yl)propoxy)propanamido)-3,3-dimethylbutanoyl)-4-hydroxy-N-(4-(4-

Precaución: for Research Use Only

Formula: C₅₉H₇₁ClFN₁₁O₇S
MW: 1132.7810 g/mol
CAS Number: 2502156-03-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR