O-(2,6-Dichlorobenzyl)hydroxylamine hydrochloride 97%

Supplier: Ambeed
Danger

A1325949-1G A1325949-250MG
76681-778EA 168.69 USD
76681-778 76681-780
O-(2,6-Dichlorobenzyl)hydroxylamine hydrochloride 97%
O-(2,6-Dichlorobenzyl)hydroxylamine hydrochloride

Precaución: for Research Use Only

Formula: C₇H₈Cl₃NO
MW: 228.5035 g/mol
MDL Number: MFCD01114588
CAS Number: 1885-52-5
UN: 1325
ADR: 4.1,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR