5-Chloro-2,3-dihydrofuro[2,3-c]pyridine 97%

Supplier: Ambeed
Warning

A1325445-250MG A1325445-1G
76681-762EA 311.91 USD
76681-762 77621-504
5-Chloro-2,3-dihydrofuro[2,3-c]pyridine 97%
5-Chloro-2,3-dihydrofuro[2,3-c]pyridine

Precaución: for Research Use Only

Formula: C₇H₆ClNO
MW: 155.5816 g/mol
CAS Number: 193605-52-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR