N-Ethylpiperazine-1-sulfonamide 97%

Supplier: Ambeed
Warning

A1325279-100MG A1325279-1G A1325279-250MG A1325279-5G
76681-750EA 68.96 USD
76681-750 76681-752 76681-754 76681-756
N-Ethylpiperazine-1-sulfonamide 97%
N-Ethylpiperazine-1-sulfonamide

Precaución: for Research Use Only

Formula: C₆H₁₅N₃O₂S
MW: 193.2672 g/mol
MDL Number: MFCD11169479
CAS Number: 1042643-73-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR