2-Amino-5-bromo-4-fluoropyridin-3-ol 97%

Supplier: Ambeed
Danger

A1324909-100MG A1324909-1G A1324909-250MG
76681-744EA 78 USD
76681-744 76681-746 76681-748
2-Amino-5-bromo-4-fluoropyridin-3-ol 97%
2-Amino-5-bromo-4-fluoropyridin-3-ol

Precaución: for Research Use Only

Formula: C₅H₄BrFN₂O
MDL Number: MFCD09835237
CAS Number: 1003710-78-8
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR