(S)-3,3'-Bis(2,4,6-triIsopropylphenyl)-1,1'-binapthyl-2,2'-diyl hydrogenphosphate 98% +99%ee

Supplier: Ambeed
Warning

Synonyms: (11bS)-4-Hydroxy-2,6-bis(2,4,6-triisopropylphenyl)dinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepine 4-oxide

A1324845-100MG A1324845-10G A1324845-1G A1324845-250MG A1324845-5G
76681-732EA 16 USD
76681-732 76681-734 76681-736 76681-738 76681-740
(S)-3,3'-Bis(2,4,6-triIsopropylphenyl)-1,1'-binapthyl-2,2'-diyl hydrogenphosphate 98% +99%ee
(S)-3,3'-Bis(2,4,6-triIsopropylphenyl)-1,1'-binapthyl-2,2'-diyl hydrogenphosphate

Precaución: for Research Use Only

Formula: C₅₀H₅₇O₄P
MDL Number: MFCD10567009
CAS Number: 874948-63-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR