4-Aminocyclohexanol mixture of isomers 98% (mixture of isomers)

Supplier: Ambeed
Danger

Synonyms: 4-Aminocyclohexan-1-ol

A1324829-1G A1324829-5G A1324829-250MG A1324829-25G
76681-722EA 17 USD
76681-722 76681-724 77867-274 77867-275
4-Aminocyclohexanol mixture of isomers 98% (mixture of isomers)
4-Aminocyclohexanol

Precaución: for Research Use Only

Formula: C₆H₁₃NO
MDL Number: MFCD00042624
CAS Number: 6850-65-3
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR