2-(p-Tolyl)piperazine dihydrochloride 97%
Supplier: Ambeed
|
Warning
|
A1324533-1G
A1324533-250MG
76681-714EA
236.59
USD
76681-714
76681-716
2-(p-Tolyl)piperazine dihydrochloride 97%
2-(p-Tolyl)piperazine dihydrochloride
Precaución: for Research Use Only
Formula:
C₁₁H₁₈Cl₂N₂ MW: 249.1800 g/mol |
CAS Number:
65709-32-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...