(2S,5S)-Hexane-2,5-diylbis(diphenylphosphine) 97%

Supplier: Ambeed
Warning

A1324491-100MG A1324491-1G A1324491-250MG
76681-708EA 42 USD
76681-708 76681-710 76681-712
(2S,5S)-Hexane-2,5-diylbis(diphenylphosphine) 97%
(2S,5S)-Hexane-2,5-diylbis(diphenylphosphine)

Precaución: for Research Use Only

Formula: C₃₀H₃₂P₂
CAS Number: 216019-41-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR