2-Hydroxy-5-methoxyisophthalaldehyde 98%

Supplier: Ambeed
Warning

A1323728-100MG A1323728-1G A1323728-250MG
76681-696EA 22 USD
76681-696 76681-698 76681-700
2-Hydroxy-5-methoxyisophthalaldehyde 98%
2-Hydroxy-5-methoxyisophthalaldehyde

Precaución: for Research Use Only

Formula: C₉H₈O₄
MDL Number: MFCD01142868
CAS Number: 73289-61-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR