(1S,2S)-N1,N1-Dimethyl-1,2-diphenylethane-1,2-diamine dihydrochloride 97%
Supplier: Ambeed
|
Warning
|
A1323484-100MG
A1323484-1G
A1323484-250MG
76681-678EA
47.75
USD
76681-678
76681-680
76681-682
(1S,2S)-N1,N1-Dimethyl-1,2-diphenylethane-1,2-diamine dihydrochloride 97%
(1S,2S)-N1,N1-Dimethyl-1,2-diphenylethane-1,2-diamine dihydrochloride
Precaución: for Research Use Only
Formula:
C₁₆H₂₂Cl₂N₂ MW: 313.2653 g/mol |
CAS Number:
2634687-77-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...