Ethyl 7-chloro-5-fluoro-1H-indole-2-carboxylate 98%

Supplier: Ambeed
Warning

A1321405-1G A1321405-5G
76681-674EA 72.15 USD
76681-674 76681-676
Ethyl 7-chloro-5-fluoro-1H-indole-2-carboxylate 98%
Ethyl 7-chloro-5-fluoro-1H-indole-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₉ClFNO₂
MW: 241.65 g/mol
MDL Number: MFCD03840761
CAS Number: 887578-55-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR