3-Bromo-2-chloro-7-iodoquinoline 98%

Supplier: Ambeed
Warning

A1321083-100MG A1321083-1G A1321083-250MG
76681-670EA 21 USD
76681-670 77566-564 77867-255
3-Bromo-2-chloro-7-iodoquinoline 98%
3-Bromo-2-chloro-7-iodoquinoline

Precaución: For research use only.

Formula: C₉H₄BrClIN
MDL Number: MFCD31801874
CAS Number: 2086768-49-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR