(2-Aminopyrimidin-4-yl)dimethylphosphine oxide 98%

Supplier: Ambeed
Warning

A1320955-100MG A1320955-1G A1320955-250MG
76681-658EA 27.59 USD
76681-658 76681-660 76681-662
(2-Aminopyrimidin-4-yl)dimethylphosphine oxide 98%
(2-Aminopyrimidin-4-yl)dimethylphosphine oxide

Precaución: for Research Use Only

Formula: C₆H₁₀N₃OP
MW: 171.1369 g/mol
MDL Number: MFCD32695665
CAS Number: 2639424-58-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR