(5-Aminopyrazin-2-yl)dimethylphosphine oxide 98%

Supplier: Ambeed
Warning

A1320924-100MG A1320924-1G A1320924-250MG A1320924-50MG
76681-650EA 135.8 USD
76681-650 76681-652 76681-654 76681-656
(5-Aminopyrazin-2-yl)dimethylphosphine oxide 98%
(5-Aminopyrazin-2-yl)dimethylphosphine oxide

Precaución: for Research Use Only

Formula: C₆H₁₀N₃OP
MW: 171.1369 g/mol
MDL Number: MFCD32695664
CAS Number: 2361977-52-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR