(R)-(+)-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic acid 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
(R)-1,2,3,4-Tetrahydro-3-isoquinolinecarboxylic Acid, H-D-Tic-OH, D-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic acid
A130899-100G
A130899-10G
A130899-1G
A130899-250MG
A130899-25G
A130899-5G
76681-626EA
140
USD
76681-626
76681-628
76681-630
76681-632
76681-634
76681-636
(R)-(+)-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic acid 98%
(R)-(+)-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic acid
Precaución: for Research Use Only
Formula:
C₁₀H₁₁NO₂ MW: 177.2 g/mol |
MDL Number:
MFCD00144038 CAS Number: 103733-65-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...