tert-Butyl 2-(2-(2-(tosyloxy)ethoxy)ethoxy)acetate 97%

Supplier: Ambeed
Warning

Synonyms: Tos-PEG2-CH2-Boc

A1282981-1G A1282981-100MG A1282981-250MG
76681-616EA 491 USD
76681-616 77867-236 77867-237
tert-Butyl 2-(2-(2-(tosyloxy)ethoxy)ethoxy)acetate 97%
tert-Butyl 2-(2-(2-(tosyloxy)ethoxy)ethoxy)acetate

Precaución: for Research Use Only

Formula: C₁₇H₂₆O₇S
MDL Number: MFCD27977515
CAS Number: 882518-89-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR