3-Chloro-4-fluoroaniline 97%, (Chunks or pellets)

Supplier: Ambeed
Danger

A1282933-1000G A1282933-25G
76681-612EA 62.6 USD
76681-612 76681-614
3-Chloro-4-fluoroaniline 97%, (Chunks or pellets)
3-Chloro-4-fluoroaniline

Precaución: for Research Use Only

Formula: C₆H₅ClFN
MW: 145.56 g/mol
MDL Number: MFCD00007767
CAS Number: 367-21-5
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR