6-Ethynylpyridin-2(1H)-one 2,2,2-trifluoroacetate 97%

Supplier: Ambeed
Warning

A1282865-100MG A1282865-1G A1282865-250MG
76681-604EA 23.34 USD
76681-604 76681-606 76681-608
6-Ethynylpyridin-2(1H)-one 2,2,2-trifluoroacetate 97%
6-Ethynylpyridin-2(1H)-one 2,2,2-trifluoroacetate

Precaución: for Research Use Only

Formula: C₉H₆F₃NO₃
MW: 233.1441 g/mol
MDL Number: MFCD32856736
CAS Number: 2682114-57-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR