(E)-3-Methyl-2-(2-nitrovinyl)thiophene 97%

Supplier: Ambeed
Warning

A1282574-1G A1282574-250MG
76681-598EA 128.37 USD
76681-598 76681-600
(E)-3-Methyl-2-(2-nitrovinyl)thiophene 97%
(E)-3-Methyl-2-(2-nitrovinyl)thiophene

Precaución: for Research Use Only

Formula: C₇H₇NO₂S
MW: 169.2010 g/mol
CAS Number: 117693-17-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR