6-Methyl-4-(trifluoromethyl)pyridazin-3-amine 97%

Supplier: Ambeed
Warning

A1282558-1G A1282558-250MG
76681-594EA 224.92 USD
76681-594 76681-596
6-Methyl-4-(trifluoromethyl)pyridazin-3-amine 97%
6-Methyl-4-(trifluoromethyl)pyridazin-3-amine

Precaución: for Research Use Only

Formula: C₆H₆F₃N₃
MW: 177.1272 g/mol
MDL Number: MFCD32711390
CAS Number: 1379378-80-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR