(R)-4-Phenyl-2-(2-((R)-p-tolylsulfinyl)phenyl)-4,5-dihydrooxazole 98%

Supplier: Ambeed
Warning

A1281596-100MG A1281596-1G A1281596-250MG
76681-576EA 169 USD
76681-576 76681-578 76681-580
(R)-4-Phenyl-2-(2-((R)-p-tolylsulfinyl)phenyl)-4,5-dihydrooxazole 98%
(R)-4-Phenyl-2-(2-((R)-p-tolylsulfinyl)phenyl)-4,5-dihydrooxazole

Precaución: for Research Use Only

Formula: C₂₂H₁₉NO₂S
CAS Number: 2828432-15-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR