(R)-2-(2-((R)-(4-(tert-Butyl)phenyl)sulfinyl)phenyl)-4-phenyl-4,5-dihydrooxazole 98%

Supplier: Ambeed
Warning

A1281592-100MG A1281592-1G A1281592-250MG A1281592-5G
76681-562EA 154 USD
76681-562 77621-476 77621-478 77867-231
(R)-2-(2-((R)-(4-(tert-Butyl)phenyl)sulfinyl)phenyl)-4-phenyl-4,5-dihydrooxazole 98%
(R)-2-(2-((R)-(4-(tert-Butyl)phenyl)sulfinyl)phenyl)-4-phenyl-4,5-dihydrooxazole

Precaución: for Research Use Only

Formula: C₂₅H₂₅NO₂S
MW: 403.5365 g/mol
CAS Number: 2828432-14-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR