(R)-2-(2-((S)-(4-(tert-Butyl)phenyl)sulfinyl)phenyl)-4-phenyl-4,5-dihydrooxazole 98%

Supplier: Ambeed
Warning

A1281589-100MG A1281589-250MG A1281589-1G A1281589-5G
76681-560EA 154 USD
76681-560 77596-702 77621-470 77867-229
(R)-2-(2-((S)-(4-(tert-Butyl)phenyl)sulfinyl)phenyl)-4-phenyl-4,5-dihydrooxazole 98%
(R)-2-(2-((S)-(4-(tert-Butyl)phenyl)sulfinyl)phenyl)-4-phenyl-4,5-dihydrooxazole

Precaución: for Research Use Only

Formula: C₂₅H₂₅NO₂S
MW: 403.5365 g/mol
CAS Number: 1965335-75-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR