(S)-4-Phenyl-2-(2-((R)-p-tolylsulfinyl)phenyl)-4,5-dihydrooxazole 95%

Supplier: Ambeed
Warning

A1281561-100MG A1281561-1G A1281561-250MG
76681-552EA 154 USD
76681-552 76681-554 76681-556
(S)-4-Phenyl-2-(2-((R)-p-tolylsulfinyl)phenyl)-4,5-dihydrooxazole 95%
(S)-4-Phenyl-2-(2-((R)-p-tolylsulfinyl)phenyl)-4,5-dihydrooxazole

Precaución: for Research Use Only

Formula: C₂₂H₁₉NO₂S
CAS Number: 396094-85-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR