4,6-Bis((R)-4-(tert-butyl)-4,5-dihydrooxazol-2-yl)dibenzo[b,d]furan 98%
Supplier: Ambeed
|
Warning
|
A1281246-100MG
A1281246-1G
A1281246-250MG
76681-522EA
148.53
USD
76681-522
76681-524
76681-526
4,6-Bis((R)-4-(tert-butyl)-4,5-dihydrooxazol-2-yl)dibenzo[b,d]furan 98%
4,6-Bis((R)-4-(tert-butyl)-4,5-dihydrooxazol-2-yl)dibenzo[b,d]furan
Precaución: for Research Use Only
Formula:
C₂₆H₃₀N₂O₃ MW: 418.5280 g/mol |
MDL Number:
MFCD34563623 CAS Number: 2757082-90-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...