2-Ethyl-5-methylpyrazine 98+% mix

Supplier: Ambeed
Warning

Synonyms: 2-Ethyl-5-methylpyrazine(contains 2-Ethyl-6-methylpyrazine)

A1281156-100G A1281156-1G A1281156-25G A1281156-5G
76681-514EA 325 USD
76681-514 76681-516 76681-518 76681-520
2-Ethyl-5-methylpyrazine 98+% mix
2-Ethyl-5-methylpyrazine

Precaución: for Research Use Only

Formula: C₇H₁₀N₂
MDL Number: MFCD00055034
CAS Number: 13360-64-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR