3-Oxo-1-phenyl-2,7,10,13-tetraoxa-4-azapentadecan-15-oic acid 97%

Supplier: Ambeed
Warning

Synonyms: Cbz-NH-PEG3-CH2COOH

A1281104-1G A1281104-250MG A1281104-5G
76681-506EA 220.67 USD
76681-506 76681-508 76681-510
3-Oxo-1-phenyl-2,7,10,13-tetraoxa-4-azapentadecan-15-oic acid 97%
3-Oxo-1-phenyl-2,7,10,13-tetraoxa-4-azapentadecan-15-oic acid

Precaución: for Research Use Only

Formula: C₁₆H₂₃NO₇
MW: 341.3563 g/mol
MDL Number: MFCD27978797
CAS Number: 462100-05-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR