4,6-Diaminopyrimidin-2(1H)-one hydrochloride 97%

Supplier: Ambeed
Warning

A1281084-1G A1281084-250MG
76681-502EA 205.82 USD
76681-502 76681-504
4,6-Diaminopyrimidin-2(1H)-one hydrochloride 97%
4,6-Diaminopyrimidin-2(1H)-one hydrochloride

Precaución: for Research Use Only

Formula: C₄H₇ClN₄O
MW: 162.5776 g/mol
MDL Number: MFCD00136695
CAS Number: 197571-63-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR