Nickel(II) meso-tetraphenylporphine 95%

Supplier: Ambeed
Warning

Synonyms: 5,10,15,20-Tetraphenylporphyrin nickel, 5,10,15,20-Tetraphenyl-21H,23H-porphine nickel(II), Nickel TPP

A1281064-1G A1281064-250MG A1281064-5G
76681-494EA 33 USD
76681-494 76681-496 76681-498
Nickel(II) meso-tetraphenylporphine 95%
Nickel(II) meso-tetraphenylporphine

Precaución: for Research Use Only

Formula: C₄₄H₂₈N₄Ni
MW: 671.41 g/mol
MDL Number: MFCD00010727
CAS Number: 14172-92-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR