2,2'-(9,9-Dioctyl-9H-fluorene-2,7-diyl)dithiophene 98%

Supplier: Ambeed
Warning

A1280618-100MG A1280618-1G A1280618-250MG
76681-488EA 29.71 USD
76681-488 76681-490 76681-492
2,2'-(9,9-Dioctyl-9H-fluorene-2,7-diyl)dithiophene 98%
2,2'-(9,9-Dioctyl-9H-fluorene-2,7-diyl)dithiophene

Precaución: for Research Use Only

Formula: C₃₇H₄₆S₂
MW: 554.8912 g/mol
MDL Number: MFCD23143975
CAS Number: 338469-45-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR