1-Bromo-8-iododibenzofuran 98%

Supplier: Ambeed
Warning

A1278492-100MG A1278492-1G A1278492-250MG A1278492-5G
76681-468EA 37.14 USD
76681-468 76681-470 76681-472 76681-474
1-Bromo-8-iododibenzofuran 98%
1-Bromo-8-iododibenzofuran

Precaución: for Research Use Only

Formula: C₁₂H₆BrIO
MW: 372.9839 g/mol
CAS Number: 1822311-11-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR