4,4' -Bis(N,N-dimethylamino)-2,2' -bipyridine 98%

Supplier: Ambeed
Warning

A1278313-100MG A1278313-1G A1278313-250MG A1278313-5G
76681-460EA 20 USD
76681-460 76681-462 76681-464 76681-466
4,4' -Bis(N,N-dimethylamino)-2,2' -bipyridine 98%
4,4' -Bis(N,N-dimethylamino)-2,2' -bipyridine

Precaución: for Research Use Only

Formula: C₁₄H₁₈N₄
MW: 242.3195 g/mol
MDL Number: MFCD00233881
CAS Number: 85698-56-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR