(R)-1-Bromo-2-methylbutane ≥98% stabilized

Supplier: Ambeed
Danger


Stabilization: Stabilised with potassium carbonate

A1276282-250MG A1276282-1G
76681-440EA 153.84 USD
76681-440 77566-476
(R)-1-Bromo-2-methylbutane ≥98% stabilized
(R)-1-Bromo-2-methylbutane

Precaución: For research use only.

Formula: C₅H₁₁Br
MW: 151.04 g/mol
MDL Number: MFCD20760246
CAS Number: 99032-67-4
UN: 2810
ADR: 6.1,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR