N2,N3-Bis(4-fluorophenyl)quinoxaline-2,3-diamine 98%
Supplier: Ambeed
|
Warning
|
A1275711-10MG
A1275711-1MG
A1275711-25MG
A1275711-50MG
A1275711-5MG
76681-422EA
152.77
USD
76681-422
76681-424
76681-426
76681-428
76681-430
N2,N3-Bis(4-fluorophenyl)quinoxaline-2,3-diamine 98%
N2,N3-Bis(4-fluorophenyl)quinoxaline-2,3-diamine
Precaución: for Research Use Only
Formula:
C₂₀H₁₄F₂N₄ MW: 348.3488 g/mol |
MDL Number:
MFCD00607076 CAS Number: 195822-23-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...