3,7-Bis(methylthio)dibenzo[b,d]thiophene 97%
Supplier: Ambeed
|
Warning
|
A1275539-1G
A1275539-250MG
76681-412EA
237.65
USD
76681-412
76681-414
3,7-Bis(methylthio)dibenzo[b,d]thiophene 97%
3,7-Bis(methylthio)dibenzo[b,d]thiophene
Precaución: for Research Use Only
Formula:
C₁₄H₁₂S₃ MW: 276.4401 g/mol |
CAS Number:
185540-18-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...