1-Chloro-3-fluoro-5-methoxy-2-nitrobenzene 97%

Supplier: Ambeed
Warning

A1275475-100MG A1275475-1G A1275475-250MG
76681-398EA 56.23 USD
76681-398 76681-400 76681-402
1-Chloro-3-fluoro-5-methoxy-2-nitrobenzene 97%
1-Chloro-3-fluoro-5-methoxy-2-nitrobenzene

Precaución: for Research Use Only

Formula: C₇H₅ClFNO₃
MW: 205.5709 g/mol
CAS Number: 2384936-24-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR