Potassium osmate(VI) dihydrate 99%

Supplier: Ambeed
Danger

Synonyms: Potassium osmium(VI) oxide dihydrate, Potassium dioxidodioxoosmium dihydrate, Dipotassium osmate dihydrate

A1275403-10G A1275403-1G A1275403-5G
76681-388EA 502.87 USD
76681-388 76681-390 76681-392
Potassium osmate(VI) dihydrate 99%
Potassium osmate(VI) dihydrate

Precaución: for Research Use Only

Formula: K₂O₄Os·2H₂O
MW: 368.45 g/mol
MDL Number: MFCD00149919
CAS Number: 10022-66-9
UN: 3085
ADR: 5.1,8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR